Order Entry
Poland
ContactUsLinkComponent
Lithium chloride ≥99% ACS, Fluka™
Irritant
Lithium chloride ≥99% ACS, Fluka™
Certificates
Irritant
Lithium chloride ≥99% ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Loss on drying (105°C, 2 h):
    Max. 1.0 %
  • Titrable Base:
    Max. 0.008 meq/g
  • Water insoluble matter:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.01 %
  • Barium (Ba):
    Max. 0.003 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.01 %
  • Sodium (Na):
    Max. 0.20 %
  • Heavy metals (ICP) ACS:
    Max. 0.002 %
  • Nitrate (NO₃):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

Related Information

Frequently Bought Together