
Lithium bromide, Sigma-Aldrich®
Catalog # SIAL229733-125G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:125 g
- Seq:3445
Specifications
About this item
:
:86.84 g/mol
Merck Index:13,05548
Boiling Pt:1265 °C (1013 hPa)
Melting Pt:550 °C
Density:3.46 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-439-8
:GHS07
CAS Number:7550-35-8
:23303-71-1
Formula:LiBr
MDL Number:MFCD00011077