Order Entry
Poland
ContactUsLinkComponent
1,2-Dichlorohexafluorocyclopentene, Sigma-Aldrich®
1,2-Dichlorohexafluorocyclopentene, Sigma-Aldrich®
Catalog # SIAL545333-25G
Supplier:  Merck
CAS Number:  
1,2-Dichlorohexafluorocyclopentene, Sigma-Aldrich®
Catalog # SIAL545333-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :244.95 g/mol
Boiling Pt:90 °C (1013 hPa)
Melting Pt:–106 °C
Density:1.651 g/cm³ (20 °C)
EINECS:211-895-4
UN:2810
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
CAS Number:706-79-6
Formula:C₅Cl₂F₆
MDL Number:MFCD00019302