3-Methyl-1,2-cyclohexanedione, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3008-43-3
:
Formula:C₇H₁₀O₂
MDL Number:MFCD00209518
:126.16 g/mol
:M-Clarity™ quality level = MQ300
EINECS:221-122-2
Frequently Bought Together