
1,3-Cyclohexadiene 96% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:592-57-4
:
Formula:C₆H₈
MDL Number:MFCD00001532
:80.13 g/mol
Boiling Pt:79…81 °C
Melting Pt:–89 °C
Density:0.847
Flash Pt:–8 °C (–144 °F)
UN:3295
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:209-764-1
:GHS02
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together