Order Entry
Poland
Orders LinkContactUsLinkComponent
p-Benzoquinone

p-Benzoquinone

Glass bottle for solids, 1 kg
Catalog # MOLE34818722-1KG
Toxic
Environment
Marketsource
Supplier:   Molekula
p-Benzoquinone
Catalog # MOLE34818722-1KG
Supplier:  Molekula
Marketsource
CAS Number:  106-51-4
p-Benzoquinone
p-Benzoquinone

Glass bottle for solids, 1 kg

CAS Number:  106-51-4
Price
1 700 zł
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone
  • CAS Number:106-51-4
  • Formula:C₆H₄O₂
  • MDL Number:MFCD00001591
  • Molecular Weight:108.1 g/mol
  • Merck Index:12,08259
  • Boiling Pt:∼174 °C (1013 hPa)
  • Melting Pt:112…114 °C
  • Density:1.318 g/cm³ (20 °C)
  • Flash Pt:77 °C
  • Storage Temperature:Ambient
  • EINECS:203-405-2
  • UN:2587
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications