p-Benzoquinone 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone
- CAS Number:106-51-4
- Formula:C₆H₄O₂
- MDL Number:MFCD00001591
- Molecular Weight:108.1 g/mol
- Merck Index:12,08259
- Boiling Pt:~ 174 °C (1013 hPa)
- Melting Pt:112…116 °C
- Density:1.31 g/cm³
- Flash Pt:77 °C
- Storage Temperature:Ambient
- EINECS:203-405-2
- UN:2587
- ADR:6.1,II
5 Options Available Below
Product Details & Documents
Synonyms: p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone
- CAS Number:106-51-4
- Formula:C₆H₄O₂
- MDL Number:MFCD00001591
- Molecular Weight:108.1 g/mol
- Merck Index:12,08259
- Boiling Pt:~ 174 °C (1013 hPa)
- Melting Pt:112…116 °C
- Density:1.31 g/cm³
- Flash Pt:77 °C
- Storage Temperature:Ambient
- EINECS:203-405-2
- UN:2587
- ADR:6.1,II
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to green Powder
- Infrared spectrum:Conforms
- Melting point:112 to 116 °C
- Titration Iodometric:≥98.5 %
- Water:≤0.5 %
- Specific optical rotation:-218° to -224° (20 °C, 589 nm) (c=2, 1M HCl) (On dried substance)
- Titration:98.5 to 101.0 % (On dried substance)
- Sulfates:≤300 ppm
- Chlorides:≤200 ppm
- Ammonium (NH4):≤200 ppm
- Iron (Fe):≤10 ppm
- Ninhydrin-positive substances:Passes test
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



