Order Entry
Poland
Orders LinkContactUsLinkComponent
Quinaldic acid, Sigma-Aldrich®

Quinaldic acid, Sigma-Aldrich®

Irritant
Supplier:   Merck
Quinaldic acid, Sigma-Aldrich®
Supplier:  Merck
Certificates
Quinaldic acid, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
  • CAS Number:93-10-7
  • Formula:C₁₀H₇NO₂
  • MDL Number:MFCD00006752
  • Molecular Weight:173.17 g/mol
  • Merck Index:12,08227
  • Melting Pt:154…157 °C
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:202-218-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    2,5 g
    Log in for availability
    Log in for availability
    1 * 2,5 g
    1 * 2,5 g - 247 zł
    247 zł
    1 * 2,5 g
       247 zł

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 620 zł
    620 zł
    1 * 10 g
       620 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.