Order Entry
Poland
ContactUsLinkComponent
1,5-Diphenylcarbazide ≥97.0% analytical reagent
Irritant
1,5-Diphenylcarbazide ≥97.0% analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
1,5-Diphenylcarbazide ≥97.0% analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 97.0 %
  • Suited for Cr6+ reagent:
    Passes test
  • Melting point:
    171 - 173 °C
  • Ignition residue (SO4):
    Max. 0.1 %
  • Insolubility in methanol:
    Max. 100 ppm

Specification Test Results

Related Information

Frequently Bought Together