Order Entry
Poland
ContactUsLinkComponent
4-Phenylaniline
4-Phenylaniline
Catalog # MOLEM65840337
Supplier:  Molekula
CAS Number:  
4-Phenylaniline
Catalog # MOLEM65840337
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-Biphenylylamine, Biphenyl-4-ylamine, 4-Aminobiphenyl
 :
 :169.23 g/mol
EINECS:202-177-1
 :GHS08
 :GHS07
CAS Number:92-67-1
Formula:C₁₂H₁₁N
MDL Number:MFCD00007879
Boiling Pt:302
Melting Pt:55
Density:1.16 g/cm³ (20 °C)
Merck Index:12,01276