Order Entry
Poland
ContactUsLinkComponent
2,3,5-Trichlorobiphenyl (PCB No. 23)
2,3,5-Trichlorobiphenyl (PCB No. 23)
Catalog # PROOCIL-PCB-23-C
CAS Number:  
2,3,5-Trichlorobiphenyl (PCB No. 23)
Catalog # PROOCIL-PCB-23-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :257.55 g/mol
CAS Number:55720-44-0
Formula:C₁₂H₇Cl₃