2,3,4',5,6-Pentachlorobiphenyl (PCB No. 117)
Catalog # PROOCIL-PCB-117-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Rodzaj opakowania:Glass bottle for solids
- Ilość:5 mg
- Seq:2210
Specifications
About this item
:
:326.44 g/mol
CAS Number:68194-11-6
Formula:C₁₂H₅Cl₅