3,3',4,5-Tetrachlorobiphenyl (PCB No. 78)
Catalog # PROOCIL-PCB-78-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:291.99 g/mol
UN:3432
ADR:9,II
CAS Number:70362-49-1
Formula:C₁₂H₆Cl₄