Order Entry
Poland
ContactUsLinkComponent
2,3,3',5,5',6-Hexachlorobiphenyl (PCB No. 165)
2,3,3',5,5',6-Hexachlorobiphenyl (PCB No. 165)
Catalog # PROOCIL-PCB-165-C
CAS Number:  
2,3,3',5,5',6-Hexachlorobiphenyl (PCB No. 165)
Catalog # PROOCIL-PCB-165-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
CAS Number:74472-46-1
Formula:C₁₂H₄Cl₆