Złóż zamówienie
Poland
ContactUsLinkComponent
2,2',3,3',5,6-Hexachlorobiphenyl (PCB No. 134)
2,2',3,3',5,6-Hexachlorobiphenyl (PCB No. 134)
Catalog # PROOCIL-PCB-134-C
CAS Number:  
2,2',3,3',5,6-Hexachlorobiphenyl (PCB No. 134)
Catalog # PROOCIL-PCB-134-C
CAS Number:  

Specifications

  • Rodzaj opakowania:
    Glass bottle for solids
  • Ilość:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
CAS Number:52704-70-8
Formula:C₁₂H₄Cl₆