Order Entry
Poland
ContactUsLinkComponent
2,5-Dichlorobiphenyl (PCB No. 9)
2,5-Dichlorobiphenyl (PCB No. 9)
Catalog # PROOCIL-PCB-9-C
CAS Number:  
2,5-Dichlorobiphenyl (PCB No. 9)
Catalog # PROOCIL-PCB-9-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :223.1 g/mol
Boiling Pt:182 °C (1013 hPa)
Melting Pt:19 °C
UN:2315
ADR:9,II
CAS Number:34883-39-1
Formula:C₁₂H₈Cl₂