Order Entry
Poland
ContactUsLinkComponent
2,2',5,5'-Tetrachlorobiphenyl (PCB No. 52)
2,2',5,5'-Tetrachlorobiphenyl (PCB No. 52)
Catalog # PROOBCR-293
CAS Number:  
2,2',5,5'-Tetrachlorobiphenyl (PCB No. 52)
Catalog # PROOBCR-293
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :291.99 g/mol
Storage Temperature:Refrigerator
UN:2315
ADR:9,II
CAS Number:35693-99-3
Formula:C₁₂H₆Cl₄