Order Entry
Poland
ContactUsLinkComponent
2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47)
2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47)
Catalog # PROOCIL-PCB-47-C
CAS Number:  
2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47)
Catalog # PROOCIL-PCB-47-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :291.99 g/mol
EINECS:219-444-3
CAS Number:2437-79-8
Formula:C₁₂H₆Cl₄