2,3',4,4',5',6-Hexachlorobiphenyl (PCB No. 168)
Catalog # PROOCIL-PCB-168-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:360.88 g/mol
CAS Number:59291-65-5
Formula:C₁₂H₄Cl₆