2,2',3,3',4,5',6'-Heptachlorobiphenyl (PCB No. 177)
Catalog # PROOCIL-PCB-177-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:395.32 g/mol
CAS Number:52663-70-4
Formula:C₁₂H₃Cl₇