2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138)
Catalog # PROOCIL-PCB-138-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:360.88 g/mol
Storage Temperature:Refrigerator
UN:2315
ADR:9,II
CAS Number:35065-28-2
Formula:C₁₂H₄Cl₆