Order Entry
Poland
ContactUsLinkComponent
 
2,2',3,4,4',5',6-Heptachlorobiphenyl (PCB No. 183)
Catalog # PROOCIL-PCB-183-C
CAS Number:  
2,2',3,4,4',5',6-Heptachlorobiphenyl (PCB No. 183)
Catalog # PROOCIL-PCB-183-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :395.32 g/mol
CAS Number:52663-69-1
Formula:C₁₂H₃Cl₇