Order Entry
Poland
ContactUsLinkComponent
2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180)
2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180)
Catalog # PROOCIL-PCB-180-C
CAS Number:  
2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180)
Catalog # PROOCIL-PCB-180-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :395.32 g/mol
UN:2315
ADR:9,II
CAS Number:35065-29-3
Formula:C₁₂H₃Cl₇