Order Entry
Poland
ContactUsLinkComponent
2,2',3,3',4,5,5',6'-Octachlorobiphenyl (PCB No. 201)
2,2',3,3',4,5,5',6'-Octachlorobiphenyl (PCB No. 201)
Catalog # PROOCIL-PCB-201-C
CAS Number:  
2,2',3,3',4,5,5',6'-Octachlorobiphenyl (PCB No. 201)
Catalog # PROOCIL-PCB-201-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :429.77 g/mol
CAS Number:52663-75-9
Formula:C₁₂H₂Cl₈