Order Entry
Poland
ContactUsLinkComponent
Triphenylmethane 99+%
Triphenylmethane 99+%
Certificates
undefined
Triphenylmethane 99+%

Specification Test Results

  • Appearance:
    Light yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    90 to 94 °C
  • GC:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together