Bromocresol green, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:76-60-8
:
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874
:698.02 g/mol
Merck Index:13,01370
Melting Pt:217…218 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-972-8
Frequently Bought Together