Bromocresol green
Catalog # MOLE34050770-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:698.02 g/mol
Merck Index:13,01370
Melting Pt:217…218 °C
Storage Temperature:Ambient
EINECS:200-972-8
CAS Number:76-60-8
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874