
o-Toluenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL557323-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.65 g/mol
Boiling Pt:272 °C (1013 hPa)
Melting Pt:10.2 °C
Density:1.3383 g/cm³ (20 °C)
EINECS:205-113-0
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
CAS Number:133-59-5
Formula:C₇H₇ClO₂S
MDL Number:MFCD00024874