2,4-Diaminotoluene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:95-80-7
:
Formula:C₇H₁₀N₂
MDL Number:MFCD00007804
:122.17 g/mol
Boiling Pt:292 °C (1013 hPa)
Melting Pt:99 °C
Density:1.045 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-453-1
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:1709
:GHS06
:GHS08
:GHS09
Frequently Bought Together