

Phenethylamine, Supelco®
numer katalogowy 41346-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:121.18 g/mol
Merck Index:12,07371
Boiling Pt:200 °C (1013 hPa)
Melting Pt:–60 °C
Density:0.9639 g/cm³ (20 °C)
Flash Pt:81 °C
EINECS:200-574-4
UN:2735
ADR:8,I
:M-Clarity™ quality level = MQ100
:GHS06
:GHS05
Storage Temperature:Ambient
CAS Number:64-04-0
Formula:C₆H₅CH₂CH₂NH₂
MDL Number:MFCD00008184