Order Entry
Poland
ContactUsLinkComponent
Phenethylamine 99%
Phenethylamine 99%
Certificates
undefined
Phenethylamine 99%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5320 to 1.5340 (20°C, 589 nm)
  • Water:
    ≤0.5 %
  • GC:
    ≥98.5 %
  • Titration with HClO4:
    98.5 to 101.5 %

Specification Test Results

Related Information

Frequently Bought Together