2-Phenylethyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2257-09-2
:
Formula:C₆H₅CH₂CH₂NCS
MDL Number:MFCD00004821
:163.24 g/mol
Density:1.09 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:218-855-5
Frequently Bought Together