2-Phenylethyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL253731-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:163.24 g/mol
Density:1.09 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:218-855-5
CAS Number:2257-09-2
Formula:C₆H₅CH₂CH₂NCS
MDL Number:MFCD00004821