Order Entry
Poland
ContactUsLinkComponent
2,6-Dinitro-4-(trifluoromethyl)aniline, Sigma-Aldrich®
Irritant
2,6-Dinitro-4-(trifluoromethyl)aniline, Sigma-Aldrich®
Catalog # SIAL559520-5G
Supplier:  Merck
CAS Number:  
Irritant
2,6-Dinitro-4-(trifluoromethyl)aniline, Sigma-Aldrich®
Catalog # SIAL559520-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :251.12 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:445-66-9
Formula:C₇H₄F₃N₃O₄
MDL Number:MFCD00024260