2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, Sigma-Aldrich®
Catalog # SIAL472190-2G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2 g
- Seq:3107
Specifications
About this item
:
Boiling Pt:53 °C (2.5 mmHg)
Density:1.418 g/cm³ (20 °C)
UN:2922
ADR:8,III
:M-Clarity™ quality level = MQ100
:205.11 g/mol
CAS Number:69922-27-6
Formula:C₈H₃F₄NO
MDL Number:MFCD00673069