
α,α,α-Trifluoro-2-tolualdehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:447-61-0
:
Formula:C₈H₅F₃O
MDL Number:MFCD00003337
:174.12 g/mol
Boiling Pt:70…71 °C (16 mmHg)
Melting Pt:–40 °C
Density:1.322
Flash Pt:61 °C (141 °F)
EINECS:207-185-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together