2',4'-Dimethoxyacetophenone, Sigma-Aldrich®
Catalog # SIALD129402-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:180.2 g/mol
EINECS:212-587-2
CAS Number:829-20-9
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00008727