Order Entry
Poland
Orders LinkContactUsLinkComponent
 

4'-Nitroacetophenone ≥98%

4'-Nitroacetophenone ≥98%
Certificates
4'-Nitroacetophenone ≥98%
4'-Nitroacetophenone ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 100-19-6 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Flash Point: 149°(300°F) Physical Form: Cryst powder Odour: Characteristic Melting Point: 77-81° Boiling Point: 201-202° Beilstein Reference: 607777 MDL No.: MFCD00007355

Synonyms:  p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
  • CAS Number:100-19-6
  • Formula:C₈H₇NO₃
  • MDL Number:MFCD00007355
  • Molecular Weight:165.15 g/mol
  • Boiling Pt:201…202 °C
  • Melting Pt:77…81 °C
  • Flash Pt:149 °C (300 °F)
  • Storage Temperature:Ambient
  • EINECS:202-827-4
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.