Order Entry
Poland
Orders LinkContactUsLinkComponent
4'-Nitroacetophenone 97%

4'-Nitroacetophenone 97%

4'-Nitroacetophenone 97%
 
4'-Nitroacetophenone 97%
4'-Nitroacetophenone 97%

 

 
 :  p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
  •  :100-19-6
  •  :C₈H₇NO₃
  •  :MFCD00007355
  •  :165.15 g/mol
  •  :202 °C (760 mmHg)
  •  :76…80 °C
  •  :149 °C
  •  :Ambient
  •  :202-827-4

Documents

 

 

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    76 to 80 °C
  • Water:
    ≤0.6 %
  • GC:
    ≥96.0 %
  • Free acid:
    ≤0.2 % (as 4-Nitrobenzoic acid)
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle for solids
    5 g
       
       
    1 * 5 g
    1 * 5 g - 181 zł
    181 zł
    1 * 5 g
       181 zł

  • Glass bottle for solids
    100 g
       
       
    1 * 100 g
    1 * 100 g - 204 zł
    204 zł
    1 * 100 g
       204 zł

  • Glass bottle
    1 kg
       
       
    1 * 1 kg
    1 * 1 kg - 918 zł
    918 zł
    1 * 1 kg
       918 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.