Order Entry
Poland
ContactUsLinkComponent
2,4-Dichlorobenzylamine, Sigma-Aldrich®
Corrosive
2,4-Dichlorobenzylamine, Sigma-Aldrich®
Catalog # SIALD58406-5G
Supplier:  Merck
CAS Number:  
Corrosive
2,4-Dichlorobenzylamine, Sigma-Aldrich®
Catalog # SIALD58406-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :176.04 g/mol
Boiling Pt:83 °C (1013 hPa)
Density:1.33 g/cm³ (20 °C)
EINECS:202-382-6
UN:2735
ADR:8,III
 :GHS05
CAS Number:95-00-1
Formula:C₇H₇Cl₂N
MDL Number:MFCD00008109