Order Entry
Poland
ContactUsLinkComponent
ω,2,4-Trichloroacetophenone, Sigma-Aldrich®
ω,2,4-Trichloroacetophenone, Sigma-Aldrich®
Catalog # SIAL159255-25G
Supplier:  Merck
CAS Number:  
ω,2,4-Trichloroacetophenone, Sigma-Aldrich®
Catalog # SIAL159255-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :223.49 g/mol
Boiling Pt:292 °C (1013 hPa)
Melting Pt:58…60 °C
Storage Temperature:Ambient
ADR:8,III
 :M-Clarity™ quality level = MQ100
EINECS:224-218-2
UN:2923
CAS Number:4252-78-2
Formula:C₈H₅Cl₃O
MDL Number:MFCD00000934