Order Entry
Poland
ContactUsLinkComponent
ω-Bromo-4-methoxyacetophenone
Irritant
ω-Bromo-4-methoxyacetophenone
Catalog # MOLE27611708-25G
Supplier:  Molekula
CAS Number:  
Irritant
ω-Bromo-4-methoxyacetophenone
Catalog # MOLE27611708-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :229.07 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:67…70 °C
Storage Temperature:Refrigerator
EINECS:220-118-8
UN:3261
ADR:8,II
 :GHS07
CAS Number:2632-13-5
Formula:C₉H₉BrO₂
MDL Number:MFCD00000201