Order Entry
Poland
Orders LinkContactUsLinkComponent
1,2-Benzenedithiol 96%

1,2-Benzenedithiol 96%

Irritant
1,2-Benzenedithiol 96%
Certificates
1,2-Benzenedithiol 96%

2 Options Available Below

Product Details & Documents
Synonyms:  Benzene-1,2-dithiol
  • CAS Number:17534-15-5
  • Formula:C₆H₆S₂
  • MDL Number:MFCD00004835
  • Molecular Weight:142.25 g/mol
  • Boiling Pt:52 - 54 °C (0,1 -rr)
  • Melting Pt:27…28 °C
  • Density:1,236 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:241-530-4

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid or solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥95 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 752 zł
    752 zł
    1 * 1 g
       752 zł

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 2 740 zł
    2 740 zł
    1 * 5 g
       2 740 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.