Pyrogallol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:87-66-1
:
Formula:C₆H₆O₃
MDL Number:MFCD00002192
:126.11 g/mol
Merck Index:13,08090
Boiling Pt:309 °C (1013 hPa)
Melting Pt:130…132 °C
Density:1.453 g/cm³ (20 °C)
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:201-762-9
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together