Pyrogallol, Sigma-Aldrich®
: Merck
:87-66-1
:
:C₆H₆O₃
:MFCD00002192
:126.11 g/mol
:13,08090
:309 °C (1013 hPa)
:130…132 °C
:1.453 g/cm³ (20 °C)
:2811
:6.1,III
:M-Clarity™ quality level = MQ200
:201-762-9
:GHS08
:GHS07
:Ambient
Frequently Bought Together