Order Entry
Poland
Orders LinkContactUsLinkComponent
Pyrocatechol, Sigma-Aldrich®

Pyrocatechol, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIAL430749-5G
Irritant
Supplier:   Merck
Pyrocatechol, Sigma-Aldrich®
Catalog # SIAL430749-5G
Supplier:  Merck
CAS Number:  120-80-9
Pyrocatechol, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  120-80-9
Price
330 zł
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,08089
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:103…105 °C
  • Density:1.0548 g/cm³ (20 °C)
  • Flash Pt:127 °C
  • Storage Temperature:Ambient
  • EINECS:204-427-5
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.