Order Entry
Poland
ContactUsLinkComponent
2,4-Dinitroaniline, Sigma-Aldrich®
Toxic
Health
Environment
2,4-Dinitroaniline, Sigma-Aldrich®
Catalog # SIALD193011-100G
Supplier:  Merck
CAS Number:  
Toxic
Health
Environment
2,4-Dinitroaniline, Sigma-Aldrich®
Catalog # SIALD193011-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :183.12 g/mol
Merck Index:13,03298
Boiling Pt:401 °C (1013 hPa)
Melting Pt:177…178 °C
Density:1.6 g/cm³ (20 °C)
Flash Pt:224 °C
Storage Temperature:Ambient
EINECS:202-553-5
UN:1596
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
 :GHS06
 :GHS08
 :GHS09
CAS Number:97-02-9
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007151