

2-Methoxy-4-nitrophenyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:190774-55-1
:
Formula:C₈H₆N₂O₃S
MDL Number:MFCD00041379
:210.21 g/mol
Melting Pt:107…109 °C
UN:2923
ADR:8,III
:GHS06
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together