

2-Nitrophenyl isothiocyanate ≥97%
Catalog # L09669.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:180.19 g/mol
Boiling Pt:135…137 °C (17 mmHg)
Melting Pt:73…76 °C
Flash Pt:>110 °C (230 °F)
UN:2923
ADR:8,III
:GHS06
:GHS05
Storage Temperature:Ambient
CAS Number:2719-30-4
Formula:C₇H₄N₂O₂S
MDL Number:MFCD00041238