
Phenyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:103-72-0
:
Formula:C₇H₅NS
MDL Number:MFCD00004798
:135.19 g/mol
Merck Index:12,07450
Boiling Pt:221 °C (1013 hPa)
Melting Pt:–21 °C
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:203-138-1
UN:2927
ADR:6.1,II
:Packaged under argon.
Flash Pt:87 °C
:GHS06
Frequently Bought Together